Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ribonuclease Inhibitor Antibody (OTI1B7), PerCP, Novus Biologicals™
SDP

Catalog No. NB273896PCP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB273896PCP 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB273896PCP Supplier Novus Biologicals Supplier No. NBP273896PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ribonuclease Inhibitor Monoclonal specifically detects Ribonuclease Inhibitor in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Ribonuclease Inhibitor
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1B7
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Placental ribonuclease inhibitor, Placental RNase inhibitor, PRI, RAIMGC4569, ribonuclease inhibitor, ribonuclease/angiogenin inhibitor, ribonuclease/angiogenin inhibitor 1MGC18200, RNHMGC54054
Gene Symbols RNH1
Host Species Mouse
Immunogen Full length recombinant protein of human Ribonuclease Inhibitor (NP_002930) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline metabolism, Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 6050.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.