Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ribophorin I Antibody (OTI3H3), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N273898A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273898A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273898A405 Supplier Novus Biologicals Supplier No. NBP273898AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ribophorin I Monoclonal specifically detects Ribophorin I in Human, Mouse, Rat, Canine, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen Ribophorin I
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI3H3
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DKFZp686B16177, dolichyl-diphosphooligosaccharide-protein glycosyltransferase 67 kDa subunit, Dolichyl-diphosphooligosaccharide--protein glycosyltransferase 67 kDa subunit, dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit 1, EC 2.4.1.119, oligosaccharyltransferase 1 homolog, OST1, ribophorin IRBPH1, ribophorin-1, RPN-I
Gene Symbols RPN1
Host Species Mouse
Immunogen Full length recombinant protein of human Ribophorin I (NP_002941) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6184.0
Target Species Human, Mouse, Rat, Canine, Monkey, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.