Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ribophorin I Antibody (OTI5B1), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB280452 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB280452 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB280452 Supplier Novus Biologicals Supplier No. NBP273905MFV450

Mouse Monoclonal Antibody

Ribophorin I Monoclonal antibody specifically detects Ribophorin I in Human, Mouse, Rat samples. It is validated for Flow Cytometry

Specifications

Antigen Ribophorin I
Applications Flow Cytometry
Classification Monoclonal
Clone OTI5B1
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias DKFZp686B16177, dolichyl-diphosphooligosaccharide-protein glycosyltransferase 67 kDa subunit, Dolichyl-diphosphooligosaccharide--protein glycosyltransferase 67 kDa subunit, dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit 1, EC 2.4.1.119, oligosaccharyltransferase 1 homolog, OST1, ribophorin IRBPH1, ribophorin-1, RPN-I
Host Species Mouse
Immunogen Full length human recombinant protein of human RPN1(NP_002941) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6184
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.