Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ribosomal Protein L17 Antibody, Novus Biologicals™
SDP

Catalog No. NBP100237 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP100237 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP100237 Supplier Novus Biologicals Supplier No. NBP100237
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Ribosomal Protein L17 Polyclonal specifically detects Ribosomal Protein L17 in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen Ribosomal Protein L17
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_000976.1
Gene Alias FLJ18762,60S ribosomal protein L23, FLJ92089, gene encoding putative NFkB activating protein, MGC117162, PD-1, ribosomal protein L17, rpL23, RPL23,60S ribosomal protein L17
Gene Symbols RPL17
Host Species Goat
Immunogen Peptide with sequence RYSLDPENPTKSC corresponding to N-Terminus according to NP_001030178.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6139
Test Specificity Both reported variants represent identical protein (NP_000976.1; NP_001030178.1)
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.