Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ribosomal protein L26 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB292506 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292506 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292506 Supplier Novus Biologicals Supplier No. NB1002130F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ribosomal protein L26 Polyclonal specifically detects Ribosomal protein L26 in Human, Mouse samples. It is validated for ChIP Assay, Immunoprecipitation, Western Blot.

Specifications

Antigen Ribosomal protein L26
Applications ChIP Assay, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate FITC
Dilution Western Blot, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP)
Formulation PBS
Gene Alias ribosomal protein L26,60S ribosomal protein L26
Host Species Rabbit
Immunogen Maps to a region between residue 1 and 50 of human Ribosomal Protein L26 using the numbering given in entry NP_000978.1 (GeneID 6154).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 6154
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.