Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ribosomal protein L26 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220161 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220161 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220161 Supplier Novus Biologicals Supplier No. NBP220161
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ribosomal protein L26 Polyclonal specifically detects Ribosomal protein L26 in Human samples. It is validated for Western Blot.

Specifications

Antigen Ribosomal protein L26
Applications Western Blot
Classification Polyclonal
Concentration 0.83mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. P61254
Gene Alias ribosomal protein L26,60S ribosomal protein L26
Gene Symbols RPL26
Host Species Rabbit
Immunogen Full length human ribosomal protein L26 Recombinant protein.
Molecular Weight of Antigen 17 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 6154
Test Specificity Human. Species reactivity based on sequence identity: Mouse (100%), Rhesus Monkey (100%), Pig (100%), Rat (100%), Bovine (100%), Canine (100%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.