Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ribosomal Protein S6/RPS6 Antibody (522731), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB368020 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB368020 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB368020 Supplier Novus Biologicals Supplier No. FAB5436U
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ribosomal Protein S6/RPS6 Monoclonal specifically detects Ribosomal Protein S6/RPS6 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.

Specifications

Antigen Ribosomal Protein S6/RPS6
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 522731
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias Phosphoprotein NP33, 40S ribosomal protein S6, pS6, ribosomal protein S6, S6
Host Species Mouse
Immunogen E. coli-derived recombinant human RPS6, Met1-Lys249, Accession # P62753
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, mTOR Pathway, Translation Control
Primary or Secondary Primary
Gene ID (Entrez) 6194
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.