Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ribosomal Protein S6/RPS6, p Ser235, p Ser236 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001553 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001553 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001553 Supplier Novus Biologicals Supplier No. NB1001553
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ribosomal Protein S6/RPS6 (p Ser235, p Ser236) Polyclonal specifically detects Ribosomal Protein S6/RPS6 (p Ser235, p Ser236) in Human samples. It is validated for Immunohistochemistry, Western Blot.

Specifications

Antigen Ribosomal Protein S6/RPS6 (p Ser235, p Ser236)
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2500-1:10000, Immunohistochemistry 1:10-1:500
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_001001.2.
Gene Alias Phosphoprotein NP33, 40S ribosomal protein S6, pS6, ribosomal protein S6, S6
Gene Symbols RPS6
Host Species Rabbit
Immunogen Immunogen for this antibody was a phosphorylated synthetic peptide, which represented a portion of human Ribosomal Protein S6 (GeneID 6194) around serine 235 and serine 236 according to the numbering given in entry NP_001001.2.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, mTOR Pathway, Phospho Specific, Translation Control
Primary or Secondary Primary
Gene ID (Entrez) 6194
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.