Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ribosome maturation protein SBDS Antibody (1E8), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP22294A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP22294A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP22294A94 Supplier Novus Biologicals Supplier No. NBP222594AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ribosome maturation protein SBDS Monoclonal specifically detects Ribosome maturation protein SBDS in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Ribosome maturation protein SBDS
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 100000000
Conjugate Alexa Fluor 594
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias FLJ10917, ribosome maturation protein SBDS, SDSCGI-97, Shwachman-Bodian-Diamond syndrome, Shwachman-Bodian-Diamond syndrome protein, SWDS
Gene Symbols SBDS
Host Species Mouse
Immunogen Recombinant human SBDS (1-250aa) purified from E. coli
Quantity 0.1 mL
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 51119
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.