Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ribosome maturation protein SBDS Antibody (1E8), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NBP22594549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP22594549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP22594549 Supplier Novus Biologicals Supplier No. NBP222594JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ribosome maturation protein SBDS Monoclonal specifically detects Ribosome maturation protein SBDS in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Ribosome maturation protein SBDS
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 100000000
Conjugate Janelia Fluor 549
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ10917, ribosome maturation protein SBDS, SDSCGI-97, Shwachman-Bodian-Diamond syndrome, Shwachman-Bodian-Diamond syndrome protein, SWDS
Gene Symbols SBDS
Host Species Mouse
Immunogen Recombinant human SBDS (1-250aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 51119
Test Specificity Anti-human SBDS mAb, clone 1E8, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human SBDS protein.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.