Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ribosome maturation protein SBDS Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB104188 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB104188 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB104188 Supplier Novus Biologicals Supplier No. NBP298463B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ribosome maturation protein SBDS Polyclonal specifically detects Ribosome maturation protein SBDS in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Ribosome maturation protein SBDS
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias FLJ10917, ribosome maturation protein SBDS, SDSCGI-97, Shwachman-Bodian-Diamond syndrome, Shwachman-Bodian-Diamond syndrome protein, SWDS
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Shwachman Bodian-Diamond syndrome (Q9Y3A5; Met1-Glu250).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 51119
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.