Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rictor Antibody (7B3), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N151645APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N151645APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N151645APC7 Supplier Novus Biologicals Supplier No. NBP151645APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rictor Monoclonal specifically detects Rictor in Human, Mouse, Primate samples. It is validated for Flow Cytometry.

Specifications

Antigen Rictor
Applications Flow Cytometry
Classification Monoclonal
Clone 7B3
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias AVO3, AVO3 homolog, hAVO3, KIAA1999DKFZp686B11164, mAVO3, MGC39830, PIA, pianissimo, rapamycin-insensitive companion of mTOR, RPTOR independent companion of MTOR, complex 2, TORC2-specific protein AVO3
Gene Symbols RICTOR
Host Species Mouse
Immunogen Purified recombinant fragment of human RICTOR expressed in E. coli. [UniProt No. Q6R327]
Purification Method Ammonium sulfate precipitation
Quantity 0.1 mL
Research Discipline Apoptosis, Autophagy, Cancer, DNA Repair, Hypoxia, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 253260.0
Target Species Human, Mouse, Primate
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.