Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rictor Antibody (7B3) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB013998 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013998 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013998 Supplier Novus Biologicals Supplier No. NBP280925
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rictor Monoclonal specifically detects Rictor in Human, Mouse, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.

Specifications

Antigen Rictor
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone 7B3
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 1:500-1:2000, Flow Cytometry 1 ug per million cells, ELISA : 1:10000, Immunohistochemistry : 1:200-1:1000, Immunocytochemistry/Immunofluorescence : 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready
Formulation PBS
Gene Alias AVO3, AVO3 homolog, hAVO3, KIAA1999DKFZp686B11164, mAVO3, MGC39830, PIA, pianissimo, rapamycin-insensitive companion of mTOR, RPTOR independent companion of MTOR, complex 2, TORC2-specific protein AVO3
Gene Symbols RICTOR
Host Species Mouse
Immunogen Purified recombinant fragment of human RICTOR expressed in E. coli. [UniProt# Q6R327]
Purification Method Ammonium sulfate precipitation
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer, DNA Repair, Hypoxia, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 253260
Test Specificity Human, mouse and monkey.
Target Species Human, Mouse, Primate
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.