Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rictor Antibody (7B3) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP151645SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP151645SS 0.025 mL
NBP151645 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP151645SS Supplier Novus Biologicals Supplier No. NBP151645SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

Rictor Monoclonal specifically detects Rictor in Human, Mouse, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Rictor
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 7B3
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1 ug per million cells, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000
Gene Accession No. Q6R327
Gene Alias AVO3, AVO3 homolog, hAVO3, KIAA1999DKFZp686B11164, mAVO3, MGC39830, PIA, pianissimo, rapamycin-insensitive companion of mTOR, RPTOR independent companion of MTOR, complex 2, TORC2-specific protein AVO3
Gene Symbols RICTOR
Host Species Mouse
Immunogen Purified recombinant fragment of human RICTOR expressed in E. coli. [UniProt# Q6R327]
Molecular Weight of Antigen 192 kDa
Purification Method Ammonium sulfate precipitation
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253260
Target Species Human, Mouse, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Precipitation
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.