Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rictor Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NB10056427SS

 View more versions of this product

Catalog No. NB10056427T


Only null left
Add to Cart

Description

Description

Rictor Polyclonal specifically detects Rictor in Human samples. It is validated for Western Blot.
Specifications

Specifications

Rictor
Polyclonal
Unconjugated
PBS containing 0.2% gelatin with 0.05% Sodium Azide
AVO3, AVO3 homolog, hAVO3, KIAA1999DKFZp686B11164, mAVO3, MGC39830, PIA, pianissimo, rapamycin-insensitive companion of mTOR, RPTOR independent companion of MTOR, complex 2, TORC2-specific protein AVO3
Rabbit
Protein G purified
RUO
Primary
Reacts with Human. Predicted to react with Mouse which has a 93% sequence identity.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Western Blot
0.5 mg/mL
Western Blot 5-7 ug/ml
Q6R327
RICTOR
A portion of amino acids 1-50 of human Rictor was used as the immunogen.
0.025 mg
Apoptosis, Autophagy, Cancer, DNA Repair, Hypoxia, mTOR Pathway
253260
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only