Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rictor Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $402.00
Specifications
Antigen | Rictor |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 5-7 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056427T
![]() |
Novus Biologicals
NB10056427SS |
0.025 mg |
Each for $181.90
|
|
|||||
NB10056427
![]() |
Novus Biologicals
NB10056427 |
0.1 mg |
Each for $402.00
|
|
|||||
Description
Rictor Polyclonal specifically detects Rictor in Human samples. It is validated for Western Blot.Specifications
Rictor | |
Western Blot 5-7 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
Q6R327 | |
253260 | |
A portion of amino acids 1-50 of human Rictor was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Autophagy, Cancer, DNA Repair, Hypoxia, mTOR Pathway | |
PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
AVO3, AVO3 homolog, hAVO3, KIAA1999DKFZp686B11164, mAVO3, MGC39830, PIA, pianissimo, rapamycin-insensitive companion of mTOR, RPTOR independent companion of MTOR, complex 2, TORC2-specific protein AVO3 | |
RICTOR | |
IgG | |
Protein G purified | |
Reacts with Human. Predicted to react with Mouse which has a 93% sequence identity. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title