Learn More
Pyramex™ Ridgeline™ Capstyle Hard Hat
Ridgeline™ Capstyle Hard Hat is strong, yet ultra-light for seemingly weightless protection.
$227.73 - $318.08
Specifications
Certifications/Compliance ANSI Z89.1 standards, Type 1 --- Class C, G, and E
For Use With (Application) Construction
Material Shell: ABS, Suspension: HDPE and high impact Nylon, Sweat band: Polyester Fabric and Polyurethane Foam
Size 6.5 to 8
Weight (Metric) 358 g
View More Specs
Products 17
Catalog Number Mfr. No. Product Type Color Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Color Price Quantity & Availability  
19-155-433
View Documents
Pyramex™
HP44110
4-Point Ratchet White
Case of 16 for $242.68
Only null left
Add to Cart
19-155-435
View Documents
Pyramex™
HP44112
4-Point Ratchet Gray
Case of 16 for $242.68
Only null left
Add to Cart
19-155-437
View Documents
Pyramex™
HP44113
4-Point Ratchet Slate Gray
Case of 16 for $236.08
Only null left
Add to Cart
19-155-443
View Documents
Pyramex™
HP44120
4-Point Ratchet Red
Case of 16 for $242.68
Only null left
Add to Cart
19-155-445
View Documents
Pyramex™
HP44130
4-Point Ratchet Yellow
Case of 16 for $231.67
Only null left
Add to Cart
19-155-447
View Documents
Pyramex™
HP44131
4-Point Ratchet Hi-vis Lime
Case of 16 for $313.80
Only null left
Add to Cart
19-155-449
View Documents
Pyramex™
HP44135
4-Point Ratchet Green
Case of 16 for $236.08
Only null left
Add to Cart
19-155-450
View Documents
Pyramex™
HP44141
4-Point Ratchet Hi-vis Orange
Case of 16 for $303.34
Only null left
Add to Cart
19-155-452
View Documents
Pyramex™
HP44160
4-Point Ratchet Blue
Case of 16 for $236.08
Only null left
Add to Cart
19-155-454
View Documents
Pyramex™
HP44162
4-Point Ratchet Light Blue
Case of 16 for $236.08
Only null left
Add to Cart
19-155-455
View Documents
Pyramex™
HP46110
6-Point Ratchet White
Case of 16 for $227.73
Only null left
Add to Cart
19-155-456
View Documents
Pyramex™
HP46120
6-Point Ratchet Red
Case of 16 for $240.38
Only null left
Add to Cart
19-155-457
View Documents
Pyramex™
HP46130
6-Point Ratchet Yellow
Case of 16 for $240.38
Only null left
Add to Cart
19-155-458
View Documents
Pyramex™
HP46131
6-Point Ratchet Hi-vis Lime
Case of 16 for $318.08
Only null left
Add to Cart
19-155-459
View Documents
Pyramex™
HP46135
6-Point Ratchet Green
Case of 16 for $245.33
Only null left
Add to Cart
19-155-460
View Documents
Pyramex™
HP46141
6-Point Ratchet Hi-vis Orange
Case of 16 for $318.08
Only null left
Add to cart
19-155-461
missing translation for 'viewDocuments'
Pyramex™
HP46160
6-Point Ratchet Blue
Case of 16 for $240.38
Only null left
Add to cart
Description
Description
  • ABS material – strong, yet ultra-light for seemingly weightless protection
  • Low profile design – offers a low center of gravity for better balance
  • Rear padded suspension – provides additional level of comfort on the wearers’ neck
  • 4-position harness points allows you to move the harness forward, backward, up or down – allowing for the most optimal position for
    ultimate comfort
  • Replaceable sweat band – padded fabric and polyurethane foam for all day wear
  • Vented pressure pad keeps the straps in place providing an additional level of comfort
  • Easy to grasp knob adjusts the suspension from 6 1/2 to 8
Specifications
Specifications
ANSI Z89.1 standards, Type 1 --- Class C, G, and E
Shell: ABS, Suspension: HDPE and high impact Nylon, Sweat band: Polyester Fabric and Polyurethane Foam
358 g
Short Brim (Cap Style)
Construction
6.5 to 8
Not Vented
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.