Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIG-I Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB092420 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB092420 100 μL
NB092421 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB092420 Supplier Novus Biologicals Supplier No. NBP303011100UL

Rabbit Polyclonal Antibody

RIG-I Polyclonal specifically detects RIG-I in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated.

Specifications

Antigen RIG-I
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias DEAD (Asp-Glu-Ala-Asp) box polypeptide 58, DEAD box protein 58, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide, DKFZp686N19181, EC 3.6.4.13, FLJ13599, probable ATP-dependent RNA helicase DDX58, retinoic acid inducible gene I, Retinoic acid-inducible gene 1 protein, Retinoic acid-inducible gene I protein, RIG-1, RIG-IDKFZp434J1111, RNA helicase RIG-I
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 830 to the C-terminus of human RIG-I (NP_055129.2).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 23586
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.