Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Rilpivirine
GREENER_CHOICE

Catalog No. p-7250127
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Non-nucleoside reverse transcriptase inhibitor

Rilpivirine is a potent diarylpyrimidine (DAPY) non-nucleoside reverse transcriptase inhibitor (NNRTI). It has high antiviral activity against wild-type HIV-1 and mutant viruses (EC50 values are 0.4 and 0.1-2.0 nM, respectively in a cell-based assay). This compound is orally bioavailable.

Specifications

Chemical Name or Material 4-[[4-[[4-[(1 E)-2-Cyanoethenyl]-2,6-dimethylphenyl]amino]-2-pyrimidinyl]amino]benzonitrile
CAS 500287-72-9
Quantity 10 mg
Target RNA/DNA Polymerase Inhibitors
Molecular Formula C22H18N6
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.