Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RILPL1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB341581 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341581 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341581 Supplier Novus Biologicals Supplier No. NBP281992MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RILPL1 Polyclonal antibody specifically detects RILPL1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen RILPL1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias FLJ39378, MGC105128, MGC99793, Rab interacting lysosomal protein-like 1, Rab-interacting lysosomal-like protein 1, RILP-like protein 1
Host Species Rabbit
Immunogen RILPL1 antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human RILPL1. The immunogen is located within the last 50 amino acids of RILPL1. Amino Acid Squence: WANTHRDDGYTEQGQEA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 353116
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.