Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RILPL2 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB341576 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341576 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341576 Supplier Novus Biologicals Supplier No. NBP281993PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RILPL2 Polyclonal antibody specifically detects RILPL2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen RILPL2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias FLJ30380, FLJ32372, MGC7036, p40phox-binding protein, Rab interacting lysosomal protein-like 2, Rab-interacting lysosomal protein-like 2, RILP-like protein 2, RLP2
Host Species Rabbit
Immunogen RILPL2 antibody was raised against an 16 amino acid synthetic peptide near the center of human RILPL2. The immunogen is located within amino acids 110 - 160 of RILPL2. Amino Acid Squence: VDLTDPNRPRFTLQEL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196383
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.