Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIMS2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB396520 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB396520 0.025 mg
NBP241077 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB396520 Supplier Novus Biologicals Supplier No. NBP2410770.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RIMS2 Polyclonal specifically detects RIMS2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen RIMS2
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Alias DKFZp781A0653, KIAA0751RIM2OBOERAB3 interacting protein 3, non-small cell lung cancer RimL3a protein, non-small cell lung cancer RimL3c protein, nuclear protein, rab3-interacting molecule 2, Rab-3-interacting molecule 2, Rab-3-interacting protein 3, RAB3IP3, regulating synaptic membrane exocytosis 2, regulating synaptic membrane exocytosis protein 2, RIM 2
Gene Symbols RIMS2
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid peptide near the carboxy terminusof human RIM2. The immunogen is located within amino acids 30 - 80 of RIM2. Amino Acid Squence: MDRQKKEEEKEQSVLK
Molecular Weight of Antigen 148 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9699
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.