Learn More
Description
Specifications
Specifications
| Antigen | RING1 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
| Classification | Monoclonal |
| Clone | 8C12F4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | EC 6.3.2, EC 6.3.2.-, Polycomb complex protein RING1, Really interesting new gene 1 protein, ring finger protein 1RING1A, RNF1E3 ubiquitin-protein ligase RING1 |
| Gene Symbols | RING1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
