Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RING1 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB290600 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290600 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290600 Supplier Novus Biologicals Supplier No. NBP204111PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RING1 Polyclonal antibody specifically detects RING1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation

Specifications

Antigen RING1
Applications Western Blot, Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation
Formulation PBS
Gene Alias EC 6.3.2, EC 6.3.2.-, Polycomb complex protein RING1, Really interesting new gene 1 protein, ring finger protein 1RING1A, RNF1E3 ubiquitin-protein ligase RING1
Host Species Rabbit
Immunogen Maps to a region between residue 150 and 200 of human Ring Finger Protein 1 using the numbering given in entry NP_002922.2 (GeneID 6015).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 6015
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.