Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
Ringers Rope Rescue Gloves
Shop All Ansell Products
            
                    Click to view available options
                
                
                    
                            
                                Color:
                            
                        
                        
                            
                                Black
                            
                        
                            
                                Red
                            
                        
                    
                            
                                Size:
                            
                        
                        
                            
                                Small
                            
                        
                            
                                Medium
                            
                        
                            
                                Large
                            
                        
                            
                                X-Large
                            
                        
                            
                                2X-Large
                            
                        
                    Specifications
Specifications
| Color | Black | 
| Material | Neoprene, Synthetic Leather | 
| Product Type | Extrication Gloves | 
| Size | X-Large | 
| For Use With (Application) | Rescue, Rappelling, and SWAT Situations | 
| Glove Cuff Style | Hook and Loop | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction