Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIOK1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP130103 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP130103 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP130103 Supplier Novus Biologicals Supplier No. NBP130103
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RIOK1 Polyclonal specifically detects RIOK1 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen RIOK1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q9BRS2
Gene Alias AD034, bA288G3.1, EC 2.7.11.1, FLJ30006, MGC12903, prematurely truncated due to insertion of intronic sequence, RIO kinase 1, RIO kinase 1 (yeast), RRP10, serine/threonine-protein kinase RIO1
Gene Symbols RIOK1
Host Species Rabbit
Immunogen maps to a region between residue 1 and 50 of human RIO kinase 1 using the numbering given in entry NP_113668.2
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 83732
Test Specificity Human. Based on 100% sequence identity, this antibody is predicted to react with Bovine, Dog, Pig, Panda, Orangutan, Gorilla, Chimpanzee, White-tufted-ear marmoset, African elephant, Small-eared galago, Little brown bat and Northern white-cheeked gibbon.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.