Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIOK2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP130099 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP130099 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP130099 Supplier Novus Biologicals Supplier No. NBP130099
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RIOK2 Polyclonal specifically detects RIOK2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Knockdown Validated.

Specifications

Antigen RIOK2
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, KnockDown
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, Immunohistochemistry, Immunoprecipitation 2-5 ug/mg lysate, Knockdown Validated
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. Q9BVS4
Gene Alias EC 2.7.11.1, FLJ11159, RIO kinase 2, RIO kinase 2 (yeast), RIO2, serine/threonine-protein kinase RIO2
Gene Symbols RIOK2
Host Species Rabbit
Immunogen Antigen maps to a region between residue 502 and 552 of human RIO kinase 2 using the numbering given in entry NP_060813.1
Molecular Weight of Antigen 63 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 55781
Test Specificity Human. Based on 100% sequence identity, this antibody is predicted to react with Bovine, Dog, Rabbit, Orangutan, Gorilla, Chimpanzee, White-tufted-ear marmoset, Naked mole rat, Little brown bat, Northern white-cheeked gibbon, and Small-eared galago.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.