Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIOK2 Antibody (OTI3E11), HRP, Novus Biologicals™
SDP

Catalog No. NBP273910H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273910H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273910H Supplier Novus Biologicals Supplier No. NBP273910H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RIOK2 Monoclonal specifically detects RIOK2 in Human, Mouse, Monkey samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen RIOK2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3E11
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias EC 2.7.11.1, FLJ11159, RIO kinase 2, RIO kinase 2 (yeast), RIO2, serine/threonine-protein kinase RIO2
Gene Symbols RIOK2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 277-552 of human RIOK2 (NP_060813) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 55781.0
Target Species Human, Mouse, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.