Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIOK3 Antibody, Novus Biologicals™
SDP

Catalog No. NB431572 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB431572 25 μL
NBP186982 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB431572 Supplier Novus Biologicals Supplier No. NBP18698225UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RIOK3 Polyclonal specifically detects RIOK3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen RIOK3
Applications Immunocytochemistry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:1000-1:2500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias DKFZp779L1370, EC 2.7.11.1, homolog of the Aspergillus nidulans sudD gene product, RIO kinase 3, RIO kinase 3 (yeast), serine/threonine-protein kinase RIO3, sudD (suppressor of bimD6, Aspergillus nidulans) homolog, sudD homolog, sudD suppressor of bimD6 homolog, SUDDsudD suppressor of Aspergillus nidulans bimD6 homolog
Gene Symbols RIOK3
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:IPQNTISCSLADVMSEQLAKELQLEEEAAVFPEVAVAEGPFITGENIDTSSDLMLAQMLQMEYDREYDAQLRREEKKFNGDSKVSISFENYRKVHPYEDSDSSEDEVDWQDTRDDPYRPAKP
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 8780
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.