Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIPK1/RIP1 Antibody (OTI2D6), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. N273911J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273911J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273911J646 Supplier Novus Biologicals Supplier No. NBP273911JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RIPK1/RIP1 Monoclonal specifically detects RIPK1/RIP1 in Human, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen RIPK1/RIP1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2D6
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Cell death protein RIP, EC 2.7.11.1, receptor (TNFRSF)-interacting serine-threonine kinase 1, receptor interacting protein, Receptor-interacting protein 1, receptor-interacting serine/threonine-protein kinase 1, RIP1, RIP-1, Rip1 kinase, RIPFLJ39204, Serine/threonine-protein kinase RIP
Gene Symbols RIPK1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 133-422 of human RIPK1/RIP1 (NP_003795) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Necroptosis
Primary or Secondary Primary
Gene ID (Entrez) 8737.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.