Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIPK2 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB022969 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB022969 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB022969 Supplier Novus Biologicals Supplier No. NBP132424H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RIPK2 Polyclonal specifically detects RIPK2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen RIPK2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias CARD3, CARD-containing IL-1 beta ICE-kinase, CARD-containing interleukin-1 beta-converting enzyme (ICE)-associated kinase, CARD-containing interleukin-1 beta-converting enzyme-associated kinase, CARDIAKCCK, EC 2.7.11, EC 2.7.11.1, growth-inhibiting gene 30, receptor interacting protein 2, receptor-interacting protein (RIP)-like interacting caspase-like apoptosisregulatory protein (CLARP) kinase, Receptor-interacting protein 2, receptor-interacting serine/threonine-protein kinase 2, receptor-interacting serine-threonine kinase 2, RICKGIG30, RIP-2, RIP2CARD-carrying kinase, RIP-like-interacting CLARP kinase
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RIP2. The exact sequence is proprietary.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8767
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.