Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIPK2 Antibody, Novus Biologicals™
SDP

Catalog No. NB431812 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB431812 25 μL
NBP186629 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB431812 Supplier Novus Biologicals Supplier No. NBP18662925UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RIPK2 Polyclonal specifically detects RIPK2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen RIPK2
Applications Immunohistochemistry (Paraffin), Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias CARD3, CARD-containing IL-1 beta ICE-kinase, CARD-containing interleukin-1 beta-converting enzyme (ICE)-associated kinase, CARD-containing interleukin-1 beta-converting enzyme-associated kinase, CARDIAKCCK, EC 2.7.11, EC 2.7.11.1, growth-inhibiting gene 30, receptor interacting protein 2, receptor-interacting protein (RIP)-like interacting caspase-like apoptosisregulatory protein (CLARP) kinase, Receptor-interacting protein 2, receptor-interacting serine/threonine-protein kinase 2, receptor-interacting serine-threonine kinase 2, RICKGIG30, RIP-2, RIP2CARD-carrying kinase, RIP-like-interacting CLARP kinase
Gene Symbols RIPK2
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:ITWEVLSRKQPFEDVTNPLQIMYSVSQGHRPVINEESLPYDIPHRARMISLIESGWAQNPDERPSFLKCLIELEPVLRTFEEITFLEAVIQLKKTKLQSVSSAIHLCDKKKMELSLNIPVNHGPQ
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8767
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.