Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIPK3/RIP3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP177299SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177299SS 0.025 mg
NBP177299 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP177299SS Supplier Novus Biologicals Supplier No. NBP177299SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 52 publications

RIPK3/RIP3 Polyclonal specifically detects RIPK3/RIP3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Gel Supershift Assay, Knockout Validated, Knockdown Validated.

Specifications

Antigen RIPK3/RIP3
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Gel Shift, Immunoassay, KnockDown
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1-0.5 ug/ml, ELISA 1:100 - 1:2000, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunoprecipitation 20 ug/mL, Immunohistochemistry-Paraffin 5 ug/ml, Gel Supershift Assay, Knockout Validated, KnockDown Validated
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. AAF03133
Gene Alias EC 2.7.11.1, Receptor Interacting Serine/Threonine Kinase 3, Receptor-interacting protein 3, receptor-interacting serine/threonine-protein kinase 3, receptor-interacting serine-threonine kinase 3, RIP3, RIP-3, RIP-like protein kinase 3
Gene Symbols RIPK3
Host Species Rabbit
Immunogen RIPK3/RIP3 Antibody was made to a 14 amino acid peptide near the carboxy terminus of murine RIP3. The immunogen is located within the last 50 amino acids of RIP3. Amino Acid Squence: AQFGRGRGWQPFHK
Molecular Weight of Antigen 53 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Immunology, Necroptosis, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11035
Test Specificity Mouse RIPK3/RIP3 Antibody has one isoform (486aa, 53 kDa). Human RIP3 has 3 isoforms, including isoform 1 (518aa, 57 kDa), isoform 2 (252aa, 28 kDa) and isoform 3 (231aa, 25 kDa). Rat RIP3 also has one isoform (478aa, 52 kDa). NBP1-77299 can detect can detect isoforms of mouse and rat as well as human isoform 1.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.