Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIPK3/RIP3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP224588SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224588SS 0.025 mg
NBP224588 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224588SS Supplier Novus Biologicals Supplier No. NBP224588SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 23 publications

RIPK3/RIP3 Polyclonal specifically detects RIPK3/RIP3 in Human, Primate, Mouse (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen RIPK3/RIP3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 3-7 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:20-1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 10 ug/ml
Formulation 0.1 ml PBS with 0.05% Sodium Azide
Gene Accession No. Q9Y572
Gene Alias EC 2.7.11.1, Receptor Interacting Serine/Threonine Kinase 3, Receptor-interacting protein 3, receptor-interacting serine/threonine-protein kinase 3, receptor-interacting serine-threonine kinase 3, RIP3, RIP-3, RIP-like protein kinase 3
Gene Symbols RIPK3
Host Species Rabbit
Immunogen RIPK3/RIP3 Antibody was made to a synthetic peptide (QEGPKDPEAWSRPQ) corresponding to amino acids 497-518 of human RIPK3/RIP3 was used as immunogen for this antibody (isoform CRA_a, 518 amino acids, NP_006862.2). This also corresponds to amino acids 298-311 of isoform CRA_b (GenBank: EAW66022.1).
Molecular Weight of Antigen 56.7 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Immunology, Necroptosis, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11035
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and chimp. This antibody is not suitable for testing for mouse RIP3.
Target Species Human, Primate, Mouse (Negative)
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.