Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIPK3/RIP3 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB216479 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB216479 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB216479 Supplier Novus Biologicals Supplier No. NBP177299C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RIPK3/RIP3 Polyclonal antibody specifically detects RIPK3/RIP3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown

Specifications

Antigen RIPK3/RIP3
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.11.1, Receptor Interacting Serine/Threonine Kinase 3, Receptor-interacting protein 3, receptor-interacting serine/threonine-protein kinase 3, receptor-interacting serine-threonine kinase 3, RIP3, RIP-3, RIP-like protein kinase 3
Host Species Rabbit
Immunogen RIPK3/RIP3 Antibody was made to a 14 amino acid peptide near the carboxy terminus of murine RIP3. The immunogen is located within the last 50 amino acids of RIP3. Amino Acid Squence: AQFGRGRGWQPFHK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Immunology, Necroptosis, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11035
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.