Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RISC Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10056648
Description
RISC Polyclonal specifically detects RISC in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
RISC | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
EC 3.4.16, HSCP1, retinoid inducible serine carboxypeptidase, retinoid-inducible serine carboxypeptidase, RISCEC 3.4.16.-, SCP1, serine carboxypeptidase 1serine carboxypeptidase 1 precursor protein | |
Rabbit | |
Protein G purified | |
RUO | |
59342 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
0.5 mg/mL | |
Western Blot 1:500-1:1000 | |
NP_067639.1 | |
SCPEP1 | |
This antibody was developed against a synthetic peptide corresponding to aa 28-43 of human RISC. | |
0.2 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction