Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RISC Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $402.00
Specifications
Antigen | RISC |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056648T
![]() |
Novus Biologicals
NB10056648SS |
0.05 mL |
Each for $181.90
|
|
|||||
NB10056648
![]() |
Novus Biologicals
NB10056648 |
0.2 mL |
Each for $402.00
|
|
|||||
Description
RISC Polyclonal specifically detects RISC in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
RISC | |
Western Blot 1:500-1:1000 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 0.02% Sodium Azide | |
EC 3.4.16, HSCP1, retinoid inducible serine carboxypeptidase, retinoid-inducible serine carboxypeptidase, RISCEC 3.4.16.-, SCP1, serine carboxypeptidase 1serine carboxypeptidase 1 precursor protein | |
SCPEP1 | |
IgG | |
Protein G purified |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
NP_067639.1 | |
59342 | |
This antibody was developed against a synthetic peptide corresponding to aa 28-43 of human RISC. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title