Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RISC Rabbit anti-Human, Alexa Fluor 700, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB103449 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB103449 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB103449 Supplier Novus Biologicals Supplier No. NBP297783AF700

Rabbit Polyclonal Antibody

RISC Polyclonal specifically detects RISC in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen RISC
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50 mM Sodium Borate
Gene Alias EC 3.4.16, HSCP1, retinoid inducible serine carboxypeptidase, retinoid-inducible serine carboxypeptidase, RISCEC 3.4.16.-, SCP1, serine carboxypeptidase 1serine carboxypeptidase 1 precursor protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RISC fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59342
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.