Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rit1 Mouse anti-Human, Mouse, Clone: 14G7, Novus Biologicals™
SDP

Catalog No. NB151739 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151739 100 μg
NB151740 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB151739 Supplier Novus Biologicals Supplier No. NBP311893100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rit1 Monoclonal antibody specifically detects Rit1 in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen Rit1
Applications Western Blot
Classification Monoclonal
Clone 14G7
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1 ug/ml
Formulation 20 mM sodium phosphate, 150 mM sodium chloride, 50% glycerol, pH 7.5
Gene Alias GTP-binding protein Rit1, GTP-binding protein Roc1, MGC125864, MGC125865, Ras-like protein expressed in many tissues, Ras-like without CAAX 1, Ras-like without CAAX protein 1, RIBBRIT, Ric-like, expressed in many tissues, RIT, ROC1Ric (Drosophila)-like, expressed in many tissues
Host Species Mouse
Immunogen Hybridoma produced by the fusion of splenocytes from mice immunized with recombinant full-length human Rit protein and mouse myeloma cells. (Uniprot: Q92963)
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 6016
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.