Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rit1 Mouse anti-Human, Mouse, Clone: 14G7, Novus Biologicals™

Mouse Monoclonal Antibody
$219.50 - $487.50
Specifications
Antigen | Rit1 |
---|---|
Clone | 14G7 |
Dilution | Western Blot 0.5 - 1 ug/ml |
Applications | Western Blot |
Classification | Monoclonal |
Description
Rit1 Monoclonal antibody specifically detects Rit1 in Human, Mouse samples. It is validated for Western BlotSpecifications
Rit1 | |
Western Blot 0.5 - 1 ug/ml | |
Monoclonal | |
Mouse | |
Signal Transduction, Vision | |
20 mM sodium phosphate, 150 mM sodium chloride, 50% glycerol, pH 7.5 | |
6016 | |
IgG1 | |
Store at -20C. Avoid freeze-thaw cycles. |
14G7 | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Mouse | |
GTP-binding protein Rit1, GTP-binding protein Roc1, MGC125864, MGC125865, Ras-like protein expressed in many tissues, Ras-like without CAAX 1, Ras-like without CAAX protein 1, RIBBRIT, Ric-like, expressed in many tissues, RIT, ROC1Ric (Drosophila)-like, expressed in many tissues | |
Hybridoma produced by the fusion of splenocytes from mice immunized with recombinant full-length human Rit protein and mouse myeloma cells. (Uniprot: Q92963) | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title