Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rit2 Antibody (OTI4A6), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP273914A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP273914A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP273914A70 Supplier Novus Biologicals Supplier No. NBP273914AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rit2 Monoclonal specifically detects Rit2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Rit2
Applications Western Blot
Classification Monoclonal
Clone OTI4A6
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias GTP-binding protein Rit2, GTP-binding protein Roc2, Ras-like protein expressed in neurons, Ras-like without CAAX 2, Ras-like without CAAX protein 2, RIBA, Ric (Drosophila)-like, expressed in neurons, Ric-like, expressed in neurons, RIN, ROC2
Gene Symbols RIT2
Host Species Mouse
Immunogen Full length recombinant protein of human Rit2 (NP_002921) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6014.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.