Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ritanserin
GREENER_CHOICESDP

Catalog No. 195550 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-555-0 50 mg
19-551-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-555-0 Supplier Tocris Bioscience™ Supplier No. 1955/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent 5-HT2 antagonist

Potent and long-acting 5-HT2 receptor antagonist (Ki = 0.39 nM). Anxiolytic in vivo.

Chemical Identifiers

CAS 87051-43-2
Molecular Formula C27H25F2N3OS

Specifications

Quantity 50 mg
Formula Weight 477.57
Purity >99%
Molecular Weight (g/mol) 477.574
Chemical Name or Material Ritanserin
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.