Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rituximab Antibody (109H9.06), Biotin, Novus Biologicals™
SDP

Catalog No. DDX5010B100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DDX5010B100 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DDX5010B100 Supplier Novus Biologicals Supplier No. DDX5010B100

Mouse Monoclonal Antibody

Rituximab Monoclonal specifically detects Rituximab in Human samples. It is validated for Western Blot.

Specifications

Antigen Rituximab
Applications Western Blot
Classification Monoclonal
Clone 109H9.06
Concentration 0.5mg/mL
Conjugate Biotin
Dilution Western Blot
Gene Alias B lymphocyte antigen CD20, B lymphocyte surface antigen B1, Bp35, CD20, LEU 16, LEU16, Membrane spanning 4 domains subfamily A member 1, MS4A1
Host Species Mouse
Immunogen Anti-Mabthera(Rituximab)
Purification Method Ion exchange chromatography
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.