Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rituximab Antibody (2260A), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB345650 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345650 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345650 Supplier Novus Biologicals Supplier No. FAB9630MFV450
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Rituximab Monoclonal specifically detects Rituximab in Multi-species samples. It is validated for Binding Inhibition, ELISA, Flow Cytometry, Western Blot.

Specifications

Antigen Rituximab
Applications Binding Inhibition Assay, ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 2260A
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Flow Cytometry, ELISA, Binding Inhibition
Formulation 50mM Sodium Borate
Gene Alias B lymphocyte antigen CD20, B lymphocyte surface antigen B1, Bp35, CD20, LEU 16, LEU16, Membrane spanning 4 domains subfamily A member 1, MS4A1
Host Species Rabbit
Immunogen Full-length Rituximab
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Multi-species
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.