Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIZ1 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB323855 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB323855 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB323855 Supplier Novus Biologicals Supplier No. NBP299748APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RIZ1 Polyclonal antibody specifically detects RIZ1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen RIZ1
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate APC
Dilution ELISA, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias EC 2.1.1.43, GATA-3 binding protein G3B, GATA-3-binding protein G3B, KMT8PR domain-containing protein 2, Lysine N-methyltransferase 8, MTB-ZFHUMHOXY1, PR domain containing 2, with ZNF domain, PR domain zinc finger protein 2, retinoblastoma protein-binding zinc finger protein, Retinoblastoma protein-interacting zinc finger protein, RIZ1, RIZ2, RIZMTE-binding protein, Zinc finger protein RIZ, zinc-finger DNA-binding protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human RIZ1 (Accession#: NP_036363.2; Met 1-Ala 200)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 7799
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.