Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RIZ1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$254.42 - $666.47
Specifications
| Antigen | RIZ1 |
|---|---|
| Dilution | Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20-1:50 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
RIZ1 Polyclonal specifically detects RIZ1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| RIZ1 | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| EC 2.1.1.43, GATA-3 binding protein G3B, GATA-3-binding protein G3B, KMT8PR domain-containing protein 2, Lysine N-methyltransferase 8, MTB-ZFHUMHOXY1, PR domain containing 2, with ZNF domain, PR domain zinc finger protein 2, retinoblastoma protein-binding zinc finger protein, Retinoblastoma protein-interacting zinc finger protein, RIZ1, RIZ2, RIZMTE-binding protein, Zinc finger protein RIZ, zinc-finger DNA-binding protein | |
| PRDM2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20-1:50 | |
| Polyclonal | |
| Rabbit | |
| Breast Cancer, Chromatin Research | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 7799 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:PPPFQYHHRNPMGIGVTATNFTTHNIPQTFTTAIRCTKCGKGVDNMPELHKHILACASASDKKRYTPKKNPVPLKQTVQPKNGVVVLDNSGKNAFRRMGQPKRLNFSVELSKMSSNKLKLNALKKKNQLVQKAI | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title