Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RKHD3 Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB165223 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165223 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB165223 Supplier Novus Biologicals Supplier No. NBP281777V
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RKHD3 Polyclonal antibody specifically detects RKHD3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen RKHD3
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias DKFZp434J0617, KIAA2009, mex-3 homolog B (C. elegans), ring finger and KH domain containing 3, RING finger and KH domain-containing protein 3, RING finger protein 195, RKHD3MGC117199, RNA-binding protein MEX3B, RNF195MEX-3B
Host Species Rabbit
Immunogen RKHD3 antibody was raised against a 16 amino acid synthetic peptide from near the center of human RKHD3. The immunogen is located within amino acids 180 - 230 of RKHD3. Amino Acid Squence: TYIVTPSRDKEPVFEV
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 84206
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.