Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RKIP Mouse anti-Human, Unconjugated, Clone: 7F12, MilliporeSigma™
Mouse Monoclonal Antibody
$192.35 - $452.71
Specifications
Antigen | RKIP |
---|---|
Clone | 7F12 |
Concentration | Please refer to lot specific datasheet. |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABC1136-25
|
MilliporeSigma
MABC113625UG |
25 μg |
Each for $192.35
|
|
|||||
MABC1136-100
|
MilliporeSigma
MABC1136100UG |
100 μg |
Each for $452.71
|
|
|||||
Description
Detects Raf kinase inhibitor protein using this mouse monoclonal Anti-RKIP, clone 7F12, Cat. No. MABC1136, tested for use in Western Blotting.Specifications
RKIP | |
Please refer to lot specific datasheet. | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Raf kinase inhibitor protein;Phosphatidylethanolamine-binding protein 1;PEBP-1;HCNPpp;Neuropolypeptide h3;Prostatic-binding protein | |
PEBP1;PBP;PEBP | |
IgG1 κ | |
Protein G purified | |
Clone 7F12 specifically detects human Raf kinase inhibitor protein, (RKIP). | |
∽23 kDa; 21.06 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
7F12 | |
Western Blot | |
Unconjugated | |
Mouse | |
Apoptosis & Cancer | |
Purified mouse monoclonal antibody IgG1 in PBS without azide. | |
NP_002558.1 | |
Purified native and denatured Raf kinase inhibitor protein (RKIP). | |
Primary | |
Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. | |
Antibodies |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title