Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RKIP/PBP Antibody (003), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB038614 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB038614 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB038614 Supplier Novus Biologicals Supplier No. NBP290295AF488
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

RKIP/PBP Monoclonal specifically detects RKIP/PBP in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen RKIP/PBP
Applications Western Blot, ELISA
Classification Monoclonal
Clone 3
Conjugate Alexa Fluor 488
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias HCNP, hippocampal cholinergic neurostimulating peptide, Neuropolypeptide h3, PBPPEBP-1, PEBPHCNPpp, phosphatidylethanolamine binding protein 1, phosphatidylethanolamine-binding protein 1, prostatic binding protein, Prostatic-binding protein, Raf kinase inhibitory protein, RKIPRaf kinase inhibitor protein
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human RKIP/PBP (NP_002558.1; Met1-Lys187).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 5037
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.