Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RLTPR Antibody (EM-53), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB002615 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002615 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002615 Supplier Novus Biologicals Supplier No. NBP262215FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RLTPR Monoclonal specifically detects RLTPR in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen RLTPR
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone EM-53
Conjugate DyLight 680
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CARMIL2, CARMIL2b, leucine rich repeat containing 16C, leucine-rich repeat-containing protein 16C, LRRC16C, RGD motif, leucine rich repeats, tropomodulin domain and proline-rich containing, RGD, leucine-rich repeat, tropomodulin and proline-rich containing protein, RGD, leucine-rich repeat, tropomodulin and proline-rich-containing protein
Gene Symbols RLTPR
Host Species Mouse
Immunogen Murine Rltpr (amino acids 1147-1397)
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 146206
Test Specificity The mouse monoclonal antibody EM-53 recognizes RLTPR/ CARMIL2, a protein playing a role in actin filament elongation.
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.